missing translation for 'onlineSavingsMsg'
Learn More

ZNF35 Monoclonal Antibody (OTI1E3), TrueMAB™, OriGene

Product Code. 17507413
Encompass_Preferred
Change view
Click to view available options
Quantity:
30 μL
100 μL
Unit Size:
100µL
30µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17507413 30 μL 30µL
17587403 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 17507413 Supplier Origene Technologies Supplier No. TA807051S

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

ZNF35 Monoclonal antibody specifically detects ZNF35 in Human samples. It is validated for Western Blot

Specifications

Antigen ZNF35
Applications Western Blot
Classification Monoclonal
Clone OTI1E3
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
Gene CITED1
Gene Accession No. Q99966
Gene Alias HF.10; HF10; mszf13; mszf20-1; mszf28; mszf68; Zfp105; Zfp239; Zfp35; Zfp-35; zinc finger protein 239; zinc finger protein 271; Zinc finger protein 35; zinc finger protein 35 (clone HF.10); zinc finger protein HF.10; Znf271; ZNF35
Host Species Mouse
Immunogen Full length human recombinant protein of human CITED1 produced in E.coli.
Purification Method Affinity chromatography
Quantity 30 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 4435
Target Species Human
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.