Learn More
Invitrogen™ ZNF354C Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5140124
Description
Gently mix before use. Optimal concentrations and conditions for each application should be determined by the user. Immunogen sequence: IEEEQEKKPL RQMIDSHEKT ISEDGNHTSL ELGKSLFTNT ALVTQQSVPI ERIPNMYYTF GKDFKQNFDL MKCFQIYPGG KPHICNECGK Highest antigen sequence indentity to the following orthologs: Rat - 54%, Mouse - 51%.
Specifications
| ZNF354C | |
| Polyclonal | |
| Unconjugated | |
| ZNF354C | |
| hKID3; KID3; Kidney, ischemia, and developmentally-regulated protein 3; KRAB-zinc finger protein synten; zinc finger protein 354C; ZNF354C | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 30832 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunocytochemistry | |
| 0.1 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q86Y25 | |
| ZNF354C | |
| Recombinant protein corresponding to Human ZNF354C. Recombinant protein control fragment (Product #RP-109162). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.