missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ZNF358 Antibody (PCRP-ZNF358-1A6) [Janelia Fluor™ 669], Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals (Bio-Techne) NBP3-14233JF669
This item is not returnable.
View return policy
Description
ZNF358 Monoclonal antibody specifically detects ZNF358 in Human samples. It is validated for Flow Cytometry,Immunoprecipitation,Western Blot,Protein Array,Immunocytochemistry/ Immunofluorescence
Specifications
| ZNF358 | |
| Monoclonal | |
| Janelia Fluor 669 | |
| hKID3, KID3Kidney, ischemia, and developmentally-regulated protein 3, KRAB-zinc finger protein synten, zinc finger protein 354C | |
| Recombinant full-length human ZNF358protein (Uniprot: Q9NW07) | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
| Flow Cytometry, Immunoprecipitation, Western Blot, Protein Array, Immunocytochemistry/Immunofluorescence | |
| PCRP-ZNF358-1A6 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| 140467 | |
| Store at 4°C in the dark. | |
| IgG2c |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction