missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ZNF365 Monoclonal Antibody (OTI4E9), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA800133
This item is not returnable.
View return policy
Description
This gene encodes several isoforms which have different expression patterns and functions. Mutation in this gene is associated with uric acid nephrolithiasis (UAN). Alternatively spliced variants, encoding distinct proteins, have been identified.Specifications
| ZNF365 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| Q70YC4 | |
| ZNF365 | |
| Human recombit protein fragment corresponding to amino acids 108-407 of human ZNF365 produced in E.coli. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
| Western Blot | |
| OTI4E9 | |
| Unconjugated | |
| ZNF365 | |
| Su48, UAN, ZNF365D | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 22891 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction