missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ZNF365 Monoclonal Antibody (OTI9H5), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA505065S
This item is not returnable.
View return policy
Description
ZNF365 Monoclonal antibody specifically detects ZNF365 in Human samples. It is validated for Immunohistochemistry (Paraffin), Western BlotSpecifications
| ZNF365 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| Q15714 | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 8848 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| IgG2a |
| Immunohistochemistry (Paraffin), Western Blot | |
| OTI9H5 | |
| Unconjugated | |
| Tsc22d1 | |
| AI839779; AV340892; DBZ; DISC1-binding zinc-finger protein; KIAA0844; LOW QUALITY PROTEIN: protein ZNF365; protein su48; Protein ZNF365; protein ZNF365-like; Su48; Talanin; UAN; Zfp365; zinc finger protein 365; ZNF365; ZNF365D | |
| Full length human recombinant protein of human TSC22D1 produced in HEK293T cell. | |
| 30 μL | |
| Primary | |
| Human | |
| Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction