missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ ZNF38 Monoclonal Antibody (OTI2E5)

Mouse Monoclonal Antibody
Brand: Invitrogen™ MA526251
This item is not returnable.
View return policy
Description
ZNF38 Monoclonal Antibody for Western Blot, ICC/IF, IHC (P)
ZSCAN21 belongs to the krueppel C2H2-type zinc-finger protein family and is a strong transcriptional activator.
Specifications
| ZNF38 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q07231, Q9Y5A6 | |
| ZSCAN21 | |
| Full length human recombinant protein of ZSCAN21 produced in HEK293T cell. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Canine, Monkey | |
| Antibody | |
| IgG1 |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| OTI2E5 | |
| Unconjugated | |
| ZSCAN21 | |
| AI326272; CTfin51; hypothetical protein LOC527103; NY-REN-21; renal carcinoma antigen NY-REN-21; RU49; Transcription factor RU49; Zfp38; zfp-38; zinc finger and SCAN domain containing 21; zinc finger and SCAN domain-containing protein 21; zinc finger and SCAN domain-containing protein 21; zinc finger protein 3; zinc finger protein with KRAB and SCAN domains 1; zinc finger proliferation 1; zinc finger protein 38; zinc finger protein 38 (KOX 25); zinc finger protein 38 homolog; zinc finger protein NY-REN-21 antigen; zinc finger with KRAB and SCAN domains 1; Zipro1; ZKSCAN1; ZNF38; ZSCAN21 | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 22697, 7589 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction