missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ZNF384 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-21002
This item is not returnable.
View return policy
Description
ZNF384 Polyclonal specifically detects ZNF384 in Human samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.
Specifications
| ZNF384 | |
| Polyclonal | |
| Unconjugated | |
| PBS (pH 7.0), 1.0% BSA and 20% Glycerol with 0.01% Thimerosal | |
| CAG repeat protein 1, CAG/CTG 2, Cas-interacting zinc finger protein, zinc finger protein 384 | |
| Rabbit | |
| 63 kDa | |
| 0.1 mL | |
| Primary | |
| Human. Species reactivity based on sequence identity: Bovine (100%). | |
| Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. |
| Western Blot, Immunocytochemistry, Immunofluorescence | |
| 1mg/mL | |
| Western Blot 1:500-1:3000, Immunocytochemistry/Immunofluorescence 1:100-1:1000 | |
| Q8TF68 | |
| ZNF384 | |
| Recombinant protein encompassing a sequence within the N-terminus region of human ZNF384. The exact sequence is proprietary. | |
| Affinity Purified | |
| RUO | |
| 171017 | |
| Human | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction