missing translation for 'onlineSavingsMsg'
Learn More

ZNF385B Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody

€ 280.00

Specifications

Antigen ZNF385B
Applications Western Blot, Immunocytochemistry, Immunofluorescence
Classification Polyclonal
Conjugate Unconjugated
Host Species Rabbit
View More Specs

Products 1
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
18643297
View Documents
Novus Biologicals
NBP2-58807-25ul
25 μL
€ 280.00
25µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

ZNF385B Polyclonal specifically detects ZNF385B in Human samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.
Specifications

Specifications

ZNF385B
Polyclonal
Rabbit
C2H2-like zinc finger protein rearranged in thyroid adenomas, DKFZp686L0787, KRAB zinc finger protein, zinc finger protein 331, Zinc finger protein 463, ZNF463RITAZNF361Zinc finger protein 361
ZNF385B
IgG
Affinity Purified
Western Blot, Immunocytochemistry, Immunofluorescence
Unconjugated
RUO
151126
This antibody was developed against a recombinant protein corresponding to the following amino acid sequence:GSKHAKKVKALDATKNKPKMVPSKDSAKANPSCSITPITGNNSDKSEDKGKLKASSSSQP
Primary
Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins.
Videos
SDS
Documents

Documents

Product Certifications

For Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.