missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ZNF385B Monoclonal Antibody (OTI1B3), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA802758S
This item is not returnable.
View return policy
Description
ZNF385B Monoclonal antibody specifically detects ZNF385B in Human samples. It is validated for Western BlotSpecifications
| ZNF385B | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| Q8TEK3 | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 84444 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| IgG1 |
| Western Blot | |
| OTI1B3 | |
| Unconjugated | |
| Dot1l | |
| B830010L13Rik; C130013B13Rik; si:busm1-42h5.1; si:by51i9.1; si:dz145b16.1; si:dz42h5.1; si:xx-51i9.1; wu:fj47d04; ZFN533; Zfp385b; Zfp533; zgc:73248; Zinc finger protein 385B; zinc finger protein 533; ZNF385B; ZNF533 | |
| Human recombinant protein fragment corresponding to amino acids 1008-1424 of human DOT1L produced in E.coli. | |
| 30 μL | |
| Primary | |
| Human | |
| Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction