Learn More
ZNF385B Mouse anti-Human, Clone: OTI6G3, lyophilized, TrueMAB™
Mouse Monoclonal Antibody
Brand: Origene Technologies CF802760
Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
ZNF385B contains 4 U1-type zinc fingers. ZNF385B is expressed in brain with a potential role in RNA maturation, stability, or transcription regulation. There are 5 known isoforms.Specifications
| ZNF385B | |
| Monoclonal | |
| Unconjugated | |
| ZNF385B | |
| ZNF533 | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 151126 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Lyophilized |
| Western Blot | |
| OTI6G3 | |
| PBS with 8% trehalose and no preservative; pH 7.3 | |
| Q569K4 | |
| ZNF385B | |
| Human recombit protein fragment corresponding to amino acids 149-399 of human ZNF385B produced in E.coli. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.