missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ZNF394 Monoclonal Antibody (OTI3E4), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA808958S
This item is not returnable.
View return policy
Description
ZNF394 Monoclonal antibody specifically detects ZNF394 in Human samples. It is validated for Western BlotSpecifications
| ZNF394 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| Q16698 | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 1666 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| IgG1 |
| Western Blot | |
| OTI3E4 | |
| Unconjugated | |
| DECR1 | |
| 2310046C23Rik; 2810437E14Rik; Rlzfy; RLZF-Y; SNAP19; truncated zinc finger protein isoform; Unknown (protein for MGC:127479); Zfp394; Zfp94; zfp-94; Zfp99; Zinc finger protein 394; zinc finger protein 94; zinc finger protein 99; zinc finger protein LOC522836; zinc finger protein with KRAB and SCAN domains 14; Zinc finger protein Y1; zinc finger protein Y1 (RLZF-Y); zinc finger with KRAB and SCAN domains 14; ZKSCAN14; Znf394; ZSCAN46 | |
| Full length human recombinant protein of human DECR1 produced in HEK293T cell. | |
| 30 μL | |
| Primary | |
| Human | |
| Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction