missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ ZNF396 Polyclonal Antibody
GREENER_CHOICE

Rabbit Polyclonal Antibody

Brand:  Invitrogen™ PA530926

Product Code. 13495427

  • € 516.00 / 100µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Recommended positive controls: 293T, A431, Jurakt, Raji. Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

Isoform 1 and isoform 2 act as DNA-dependent transcriptional repressors.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

ZNF396
Polyclonal
Unconjugated
ZNF396
Zinc finger and SCAN domain-containing protein 14; zinc finger protein 396; ZNF396; ZSCAN14
Rabbit
Antigen affinity chromatography
RUO
252884
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Western Blot
1 mg/mL
PBS with 1% BSA, 20% glycerol and 0.01% thimerosal; pH 7
Q96N95
ZNF396
Recombinant protein encompassing a sequence within the center region of human ZNF396. The exact sequence is proprietary.
100 μL
Primary
Human
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.