missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ZNF397 Antibody (OTI1F3), Janelia Fluor™ 549, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-74950JF549
This item is not returnable.
View return policy
Description
ZNF397 Monoclonal specifically detects ZNF397 in Human, Rat samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.
Specifications
| ZNF397 | |
| Monoclonal | |
| Janelia Fluor 549 | |
| 50mM Sodium Borate with 0.05% Sodium Azide | |
| ZNF397 | |
| Full length recombinant protein of human ZNF397 (NP_115723) produced in HEK293T cells. | |
| 0.1 mL | |
| 84307.0 | |
| Store at 4°C in the dark. |
| Western Blot, Immunocytochemistry, Immunofluorescence | |
| OTI1F3 | |
| Western Blot, Immunocytochemistry/Immunofluorescence | |
| KIAA1588, KRAB-containing zinc finger protein 317, zinc finger protein 317 | |
| Mouse | |
| Affinity Purified | |
| Primary | |
| Human, Rat | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction