missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ZNF398 Antibody - Azide and BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-94651-0.02ml
This item is not returnable.
View return policy
Description
ZNF398 Polyclonal antibody specifically detects ZNF398 in Human, Mouse, Rat samples. It is validated for Western Blot
Specifications
| ZNF398 | |
| Polyclonal | |
| Western Blot 1:1000 - 1:5000 | |
| KIAA0961zinc finger protein 30 homolog, zfp-30, zinc finger protein 30 homolog (mouse), Zinc finger protein 745, ZNF745Zfp-30 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 130-230 of human ZNF398 (NP_733787.1). LPPGIKGDIPKVPVAFDDVSIYFSTPEWEKLEEWQKELYKNIMKGNYESLISMDYAINQPDVLSQIQPEGEHNTEDQAGPEESEIPTDPSEEPGISTSDIL | |
| 0.02 mL | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
| Western Blot | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| 57541 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction