missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ ZNF398 Polyclonal Antibody
GREENER_CHOICE

Rabbit Polyclonal Antibody

Brand:  Invitrogen™ PA589431

Product Code. 16577331

  • € 520.00 / 100µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Immunogen sequence: LPPGIKGDIP KVPVAFDDVS IYFSTPEWEK LEEWQKELYK NIMKGNYESL ISMDYAINQP DVLSQIQPEG EHNTEDQAGP EESEIPTDPS EEPGISTSDI L; Positive Samples: HeLa, 293T, Mouse testis, Mouse brain, Rat testis; Cellular Location: Nucleus.

Function as a transcriptional activator.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

ZNF398
Polyclonal
Unconjugated
ZNF398
KIAA1339; P51; P71; ZER6; zinc finger DNA binding protein p52/p71; zinc finger DNA binding protein ZER6; zinc finger DNA-binding protein p52/p71; Zinc finger protein 398; zinc finger-estrogen receptor interaction, clone 6; ZNF398
Rabbit
Affinity chromatography
RUO
57541
-20°C, Avoid Freeze/Thaw Cycles
Liquid
ELISA, Western Blot
1.06 mg/mL
PBS with 50% glycerol and 0.01% thimerosal; pH 7.3
Q8TD17
ZNF398
Recombinant fusion protein containing a sequence corresponding to amino acids 130-230 of human ZNF398 (NP_7337871).
100 μL
Primary
Human, Mouse, Rat
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.