missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ZNF407 Antibody, Alexa Fluor™ 750, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-34852AF750
This item is not returnable.
View return policy
Description
ZNF407 Polyclonal antibody specifically detects ZNF407 in Human, Mouse samples. It is validated for Western Blot
Specifications
| ZNF407 | |
| Polyclonal | |
| Western Blot | |
| FLJ13839, FLJ20307, KIAA1703, zinc finger protein 407 | |
| The immunogen this antibody was made to, maps to a region between residue 2198 to 2248 of human Zinc Finger Protein 407. | |
| 0.1 mL | |
| Primary | |
| Human, Mouse | |
| Purified |
| Western Blot | |
| Alexa Fluor 750 | |
| 50mM Sodium Borate | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| 55628 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction