missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ZNF407 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-06058-50ul
This item is not returnable.
View return policy
Description
ZNF407 Polyclonal antibody specifically detects ZNF407 in Human samples. It is validated for Immunocytochemistry/ Immunofluorescence
Specifications
| ZNF407 | |
| Polyclonal | |
| Immunocytochemistry/ Immunofluorescence 1:100-1:500 | |
| FLJ13839, FLJ20307, KIAA1703, zinc finger protein 407 | |
| Produced in rabbits immunized with E. coli-derived Human ZNF407 fragment. | |
| 50 μg | |
| Primary | |
| Human | |
| Purified |
| Immunofluorescence | |
| Unconjugated | |
| PBS, pH7.0 | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| 55628 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction