missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ZNF408 Antibody (PCRP-ZNF408-1E5), Alexa Fluor™ 647, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-08324AF647
This item is not returnable.
View return policy
Description
ZNF408 Monoclonal antibody specifically detects ZNF408 in Human samples. It is validated for Western Blot, Flow Cytometry, Protein Array
Specifications
| ZNF408 | |
| Monoclonal | |
| Alexa Fluor 647 | |
| A8-51, C2-H2 type zinc finger protein, FLJ20216, HF.12, HZF3.1, KOX25zinc finger protein 3 (A8-51), PP838, Zfp113, zinc finger protein 3, Zinc finger protein HF.12, Zinc finger protein HZF3.1, Zinc finger protein KOX25 | |
| Recombinant full-length human ZNF408 protein (Uniprot: Q9H9D4) | |
| 100 μg | |
| Primary | |
| Human | |
| Purified |
| Western Blot, Flow Cytometry, Protein Array | |
| PCRP-ZNF408-1E5 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| 79797 | |
| Store at 4°C in the dark. | |
| IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction