missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ ZNF408 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA530826
This item is not returnable.
View return policy
Description
Recommended positive controls: H1299, H1299 nuclear extract. Predicted reactivity: Bovine (94%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
Zinc-finger proteins contain DNA-binding domains and have a wide variety of functions, most of which encompass some form of transcriptional activation or repression. The majority of zinc-finger proteins contain a Kruppel-type DNA binding domain and a KRAB domain, which is thought to interact with KAP1, thereby recruiting histone modifying proteins. As a member of the Kruppel C2H2-type zinc-finger protein family, ZNF396 (zinc finger protein 396), also known as PRDM17 (PR domain zinc finger protein 17), is a 720 amino acid nuclear protein that contains 10 C2H2-type zinc fingers. The gene encoding ZNF408 maps to human chromosome 11, which houses over 1,400 genes and comprises nearly 4% of the human genome. Jervell and Lange-Nielsen syndrome, Jacobsen syndrome, Niemann-Pick disease, hereditary angioedema and Smith-Lemli-Opitz syndrome are associated with defects in genes that maps to chromosome 11.
Specifications
| ZNF408 | |
| Polyclonal | |
| Unconjugated | |
| ZNF408 | |
| EVR6; LOC100362202; PFM14; PR domain zinc finger protein 17; PRDM17; RP72; Zinc finger protein 408; ZNF408 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 79797 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Western Blot, Immunocytochemistry | |
| 1 mg/mL | |
| PBS with 1% BSA, 20% glycerol and 0.01% thimerosal; pH 7 | |
| Q9H9D4 | |
| ZNF408 | |
| Recombinant protein encompassing a sequence within the center region of human ZNF408. The exact sequence is proprietary. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction