missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ZNF419A Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-21013
This item is not returnable.
View return policy
Description
ZNF419A Polyclonal specifically detects ZNF419A in Human samples. It is validated for Western Blot.
Specifications
| ZNF419A | |
| Polyclonal | |
| Unconjugated | |
| 0.1M Tris (pH 7.0), 0.1M Glycine and 20% Glycerol with 0.01% Thimerosal | |
| zinc finger protein 419 | |
| Rabbit | |
| 59 kDa | |
| 0.1 mL | |
| Primary | |
| Human | |
| IgG |
| Western Blot | |
| 1mg/mL | |
| Western Blot 1:500-1:3000 | |
| Q96HQ0 | |
| ZNF419 | |
| Recombinant protein encompassing a sequence within the N-terminus region of human ZNF419. The exact sequence is proprietary. | |
| Affinity Purified | |
| RUO | |
| 79744 | |
| Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction