Learn More
Invitrogen™ ZNF441 Polyclonal Antibody
Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA541218
This item is not returnable.
View return policy
Description
Peptide sequence: MVERACEIKD NSQCGGPFTQ TQDSIVNEKI PGVDPWESSE CTDVLMGRSS Sequence homology: Human: 100%.
ZNF441 contains 19 C2H2-type zinc fingers and belongs to the krueppel C2H2-type zinc-finger protein family. It may be involved in transcriptional regulation.
Specifications
| ZNF441 | |
| Polyclonal | |
| Unconjugated | |
| ZNF441 | |
| Zinc finger protein 441; ZNF441 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 126068 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide | |
| Q8N8Z8 | |
| ZNF441 | |
| Synthetic peptide directed towards the N-terminal of human ZNF441 (aa 68-117). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |