missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ZNF444 Antibody, Alexa Fluor™ 647, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-78190AF647
This item is not returnable.
View return policy
Description
ZNF444 Polyclonal antibody specifically detects ZNF444 in Human samples. It is validated for Western Blot
Specifications
| ZNF444 | |
| Polyclonal | |
| Western Blot | |
| HZF3, MGC126498, zinc finger protein 268, zinc finger protein 3, Zinc finger protein HZF3 | |
| The immunogen for this product maps to a region between residue 150 and 200 of human Zinc Finger Protein 444 using the numbering given in entry NP_060807.2 (GeneID 55311). | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
| Western Blot | |
| Alexa Fluor 647 | |
| 50mM Sodium Borate | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| 55311 | |
| Store at 4C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction