missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ZNF444 Antibody (PCRP-ZNF444-1E11), Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-23635-100ug
This item is not returnable.
View return policy
Description
ZNF444 Monoclonal antibody specifically detects ZNF444 in Human samples. It is validated for Flow Cytometry, Immunocytochemistry/ Immunofluorescence
Specifications
| ZNF444 | |
| Monoclonal | |
| 0.2 mg/mL | |
| Flow Cytometry : 1-2 μg/million cells, Immunocytochemistry/ Immunofluorescence : 1-2 μg/mL | |
| HZF3, MGC126498, zinc finger protein 268, zinc finger protein 3, Zinc finger protein HZF3 | |
| Recombinant fragment (around aa25-105) of human ZNF444 protein | |
| 100 μg | |
| Primary | |
| Human | |
| Purified |
| Flow Cytometry, Immunofluorescence | |
| PCRP-ZNF444-1E11 | |
| Unconjugated | |
| 10 mM PBS with 0.05% BSA | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| 55311 | |
| Store at 4°C. | |
| IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction