missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ ZNF445 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA568900
This item is not returnable.
View return policy
Description
This target displays homology in the following species: Mouse: 100%.
ZNF445 gene ontology annotations related to this gene include DNA-binding transcription factor activity; metal ion binding; nucleic acid binding.
Specifications
| ZNF445 | |
| Polyclonal | |
| Unconjugated | |
| Zfp445 | |
| AW610627; mszf29; mszf50; RGD1559144; Zfp445; Zinc finger protein 168; zinc finger protein 445; Zinc finger protein with KRAB and SCAN domains 15; ZKSCAN15; ZNF168; Znf445; ZSCAN47 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 235682 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide | |
| Q8R2V3 | |
| Zfp445 | |
| synthetic peptide corresponding to a region of mouse ZNF445 (aa 601-650). | |
| 100 μL | |
| Primary | |
| Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction