missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ ZNF447 Polyclonal Antibody
Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA540694
This item is not returnable.
View return policy
Description
Peptide sequence: GDALADPPSG TTEEEEEQPG KAPDPQDPQD AESDSATGSQ RQSVIQQPAP Sequence homology: Human: 100%.
May be involved in transcriptional regulation.
Specifications
| ZNF447 | |
| Polyclonal | |
| Unconjugated | |
| ZSCAN18 | |
| EG232875; RGD1560671; similar to hypothetical protein FLJ12895; zinc finger and SCAN domain containing 18; zinc finger and SCAN domain-containing protein 18; Zinc finger protein 447; ZNF447; ZSCAN18 | |
| Rabbit | |
| Protein A | |
| RUO | |
| 65982 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| 1 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide | |
| Q8TBC5 | |
| ZSCAN18 | |
| Synthetic peptide directed towards the middle region of human ZNF447. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction