missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ZNF449 Antibody (OTI4C6), HRP, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-74952H
This item is not returnable.
View return policy
Description
ZNF449 Monoclonal specifically detects ZNF449 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry.
Specifications
| ZNF449 | |
| Monoclonal | |
| HRP | |
| PBS with No Preservative | |
| ZNF449 | |
| Full length recombinant protein of human ZNF449 (NP_689908) produced in E.coli. | |
| 0.1 mL | |
| 203523.0 | |
| Store at 4°C in the dark. |
| Western Blot, Immunohistochemistry | |
| OTI4C6 | |
| Western Blot, Immunohistochemistry | |
| HZF1Zinc finger protein HZF1, KIAA2007, zinc finger protein 1, zinc finger protein 266 | |
| Mouse | |
| Affinity Purified | |
| Primary | |
| Human, Mouse, Rat | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction