missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ZNF449 Monoclonal Antibody (OTI5F6), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA808536
This item is not returnable.
View return policy
Description
This gene encodes a nuclear protein that likely functions as a transcription factor. The protein includes an N-terminal SCAN domain, and seven C2H2-type zinc finger motifs.Specifications
| ZNF449 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| Q6P9G9 | |
| ZNF449 | |
| Full length human recombit protein of human ZNF449 produced in E.coli. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
| Western Blot | |
| OTI5F6 | |
| Unconjugated | |
| ZNF449 | |
| ZSCAN19 | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 203523 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction