missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ZNF454 Monoclonal Antibody (OTI2G4), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA810948
This item is not returnable.
View return policy
Description
ZNF454 belongs to the krueppel C2H2-type zinc-finger protein family. It contains 12 C2H2-type zinc fingers and 1 KRAB domain. It may be involved in transcriptional regulation.Specifications
| ZNF454 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| Q8N9F8 | |
| ZNF454 | |
| Full length human recombit protein of human ZNF454 produced in E.coli. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG2a |
| Western Blot | |
| OTI2G4 | |
| Unconjugated | |
| ZNF454 | |
| zinc finger protein 454 | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 285676 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction