Specifications
| Quantity | 0.1 mL |
| Content And Storage | Store at -80°C. Avoid freeze/thaw cycles. |
| Target | ZNF483 |
| Synonym | BMZF-2 zinc finger 2, bone marrow, BMZF2zinc finger protein 27 (KOX 22), Bone marrow zinc finger 2, KO x 22 Zinc finger protein 233, zinc finger protein 224, Zinc finger protein 255 Zinc finger protein KO x 22, Zinc finger protein 27, ZNF255zinc finger protein ZNF255, ZNF27ZNF233 |
| Gene Symbol | ZNF483 |