missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ ZNF488 Antibody (PCRP-ZNF488-2D8) [DyLight 650], Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals™ NBP3-27049C
This item is not returnable.
View return policy
Description
ZNF488 Monoclonal antibody specifically detects ZNF488 in Human samples. It is validated for Flow Cytometry,Immunohistochemistry (Paraffin)
Specifications
| ZNF488 | |
| Monoclonal | |
| DyLight 650 | |
| FDZF2Zinc finger protein FDZF2, zinc finger protein 230 | |
| Recombinant fragment (around aa170-307) of human ZNF488 | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
| Flow Cytometry, Immunohistochemistry (Paraffin) | |
| PCRP-ZNF488-2D8 | |
| 50 mM Sodium Borate | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| 118738 | |
| Store at 4°C in the dark. | |
| IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction