missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ZNF496, Mouse anti-Human, Clone: 2B3, Abnova™
Mouse Monoclonal Antibody
Brand: Abnova H00084838-M08.100ug
This item is not returnable.
View return policy
Description
Sequence: HLQPDRLQPVEKREQAASEDADKGPKEPLENGKAKLSFQCCECGKAFQRHDHLARHRSHFHLKDKARPFQCRYCVKSFTQNYDLLRHERLHMKRRSKQALNSpecifications
| ZNF496 | |
| Monoclonal | |
| Unconjugated | |
| zinc finger protein 496 | |
| MGC15548/NIZP1/ZKSCAN17 | |
| Mouse | |
| 100 μg | |
| Primary | |
| Human, Mouse | |
| Antibody |
| ELISA, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
| 2B3 | |
| In 1x PBS, pH 7.4 | |
| NM_032752 | |
| ZNF496 | |
| ZNF496 (NP_116141, 485 a.a. ∼ 585 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. | |
| RUO | |
| 84838 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
| IgG2a κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction