missing translation for 'onlineSavingsMsg'
Learn More

ZNF496 Polyclonal Antibody
GREENER_CHOICE

Product Code. 16699794
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16699794 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16699794 Supplier Invitrogen Supplier No. PA5103348

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

ZNF496 Polyclonal Antibody

ZNF496 Polyclonal Antibody for Western Blot, ICC/IF

TRUSTED_SUSTAINABILITY

Specifications

Antigen ZNF496
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene ZNF496
Gene Accession No. Q5SXI5, Q96IT1
Gene Alias MGC15548; NIZP1; Nizp11; NSD1 (nuclear receptor binding SET-domain containing 1)-interacting zinc finger protein 1; NSD1-interacting zinc finger protein 1; Zfp496; zinc finger protein 496; Zinc finger protein with KRAB and SCAN domains 17; zinc finger with KRAB and SCAN domains 17; Zkscan17; ZNF496; ZSCAN49
Gene Symbols Zkscan17, ZNF496
Host Species Rabbit
Immunogen A synthesized peptide derived from human ZNF496(Accession Q96IT1), corresponding to amino acid residues L5-G55.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 268417, 84838
Target Species Human, Mouse
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.