missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ZNF500 Antibody (OTI2D2), DyLight 755, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-74953IR
This item is not returnable.
View return policy
Description
ZNF500 Monoclonal specifically detects ZNF500 in Human samples. It is validated for Western Blot.
Specifications
| ZNF500 | |
| Monoclonal | |
| DyLight 755 | |
| 50mM Sodium Borate with 0.05% Sodium Azide | |
| ZNF500 | |
| Recombinant protein fragment corresponding to amino acids 102-325 of human ZNF500 (NP_067678) produced in E.coli. | |
| 0.1 mL | |
| 26048.0 | |
| Store at 4°C in the dark. |
| Western Blot | |
| OTI2D2 | |
| Western Blot | |
| C2H2-171, RP58Zinc finger and BTB domain-containing protein 18, TAZ1, TAZ-1Zinc finger protein C2H2-171, Transcriptional repressor RP58, Translin-associated zinc finger protein 1, ZBTB1858 kDa repressor protein, zinc finger protein 238 | |
| Mouse | |
| Affinity Purified | |
| Primary | |
| Human | |
| IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction