missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ZNF503 Antibody, Alexa Fluor™ 488, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-32132AF488
This item is not returnable.
View return policy
Description
ZNF503 Polyclonal antibody specifically detects ZNF503 in Human samples. It is validated for Western Blot
Specifications
| ZNF503 | |
| Polyclonal | |
| Western Blot | |
| KOX17Zinc finger protein 191, lacks zinc finger domain, Retinoic acid suppression protein A, RSG-A, Zfp191, Zinc finger and SCAN domain-containing protein 3, zinc finger protein 24, zinc finger protein 24 (KOX 17), ZNF191, ZSCAN3Zinc finger protein KOX17 | |
| The immunogen this antibody was made to, maps to a region between residue 275 to 325 of human Zinc Finger Protein 503 using the numbering given in entry NP_116161.2 (GeneID 84858). | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
| Western Blot | |
| Alexa Fluor 488 | |
| 50mM Sodium Borate | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| 84858 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction