missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ZNF558 Monoclonal Antibody (OTI2C4), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA811169
This item is not returnable.
View return policy
Description
ZNF558 may be involved in transcriptional regulation.Specifications
| ZNF558 | |
| Monoclonal | |
| Unconjugated | |
| ZNF558 | |
| zinc finger protein 558 | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 148156 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| OTI2C4 | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| Q96NG5 | |
| ZNF558 | |
| Full length human recombit protein of human ZNF558 produced in E.coli. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG2b |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction