missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ ZNF568 Polyclonal Antibody
GREENER_CHOICE

Rabbit Polyclonal Antibody

Brand:  Invitrogen™ PA5139778

Product Code. 17894290

  • € 464.00 / 100µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Gently mix before use. Optimal concentrations and conditions for each application should be determined by the user. Immunogen sequence: EPWVMEEEMF GRHCPEVWEV DEQIKKQQET LVRKVTSISK KILIKEKVIE CKKVAKIFPL SSDIVTSRQS FYDCDSLDKG LEHNLDLLRY EKGCVREKQS NEFGKPFYHC ASYVVT Highest antigen sequence indentity to the following orthologs: Rat - 26%, Mouse - 25%.

ZNF568 gene ontology annotations related to this gene include regulation of transcription, DNA-templated.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

ZNF568
Polyclonal
Unconjugated
ZNF568
LOC100061744; LOW QUALITY PROTEIN: zinc finger protein 568; zinc finger protein 568; ZNF568
Rabbit
Antigen affinity chromatography
RUO
374900
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Immunocytochemistry
0.05 mg/mL
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Q3ZCX4
ZNF568
Recombinant protein corresponding to Human ZNF568. Recombinant protein control fragment (Product #RP-108755).
100 μL
Primary
Human
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.