missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ ZNF578 Polyclonal Antibody
GREENER_CHOICE

Product Code. 15596112
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15596112 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15596112 Supplier Invitrogen™ Supplier No. PA543573

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Peptide sequence: EKDIHDFEFQ SQKDERNGHE ASMPKIKELM GSTDRHDQRH AGNKPIKDQL Sequence homology: Human: 100%.

The specific function of ZNF578 is not yet known.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ZNF578
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene ZNF578
Gene Accession No. Q96N58
Gene Alias Zinc finger protein 578; ZNF578
Gene Symbols ZNF578
Host Species Rabbit
Immunogen Synthetic peptide directed towards the N-terminal of human ZNF578 (aa 107-156).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 147660
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.