missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ZNF583 Monoclonal Antibody (OTI4D2)

ZNF583 Monoclonal Antibody (OTI4D2)
Brand: Invitrogen MA527319
This item is not returnable.
View return policy
Description
Specifications
| ZNF583 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q96ND8 | |
| ZNF583 | |
| Full length human recombinant protein of ZNF583 produced in E.coli. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
| Immunohistochemistry (Paraffin), Western Blot | |
| OTI4D2 | |
| Unconjugated | |
| ZNF583 | |
| BC056221; LOC101788406; LOW QUALITY PROTEIN: zinc finger protein 583; RGD1562044; Zfp583; Zinc finger protein 583; zinc finger protein L3-5; Znf583 | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 147949 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction