missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ZNF587 Antibody (3A3), Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals H00084914-M01
This item is not returnable.
View return policy
Description
ZNF587 Monoclonal antibody specifically detects ZNF587 in Human samples. It is validated for ELISA, Immunocytochemistry/ Immunofluorescence, Sandwich ELISA
Specifications
| ZNF587 | |
| Monoclonal | |
| Unconjugated | |
| AAH17219 | |
| Mouse | |
| IgG purified | |
| RUO | |
| 84914 | |
| Aliquot and store at -20°C or -80°C. Avoid freeze-thaw cycles. | |
| IgG2a κ |
| ELISA, Immunocytochemistry/Immunofluorescence, Sandwich ELISA | |
| 3A3 | |
| In 1x PBS, pH 7.4 | |
| FLJ14710, FLJ20813, ZF6, zinc finger protein 587, zinc finger protein zfp6 | |
| ZNF587 (AAH17219, 1 a.a. ∽ 122 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. MAVSSQQGEIMESRIFFQGSHAHFPTCMNVDTAATVLAVNVNLASNHCSQGNVPIRRRLSGTLILTGRWDILRDPEAGCHLLNFPEGCLESVSSHSELFFLLWLTKNMEPHKVHCNSFIFVK | |
| 0.1 mg | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction