Learn More
ZNF594 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-68759
This item is not returnable.
View return policy
Description
ZNF594 Polyclonal antibody specifically detects ZNF594 in Human samples. It is validated for Immunocytochemistry/ Immunofluorescence
Specifications
| ZNF594 | |
| Polyclonal | |
| Immunocytochemistry/ Immunofluorescence 0.25-2 μg/mL | |
| KIAA1871, Zinc Finger Protein 594, Zinc Finger Protein HZF18 | |
| This antibody was developed against a recombinant protein corresponding to amino acids: IREMHIIPQKAIVGEIGHGCNEGEKILSAGESSHRYEVSGQNFKQKSGLTEHQK | |
| 100 μg | |
| Primary | |
| Human | |
| Purified |
| Immunofluorescence | |
| Unconjugated | |
| PBS (pH 7.2) and 40% Glycerol | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| 84622 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |