missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ ZNF594 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA566901
This item is not returnable.
View return policy
Description
Immunogen sequence: QRLHAGEKLEE CEKTFSKDEE LREEQRIHQE EKAYWCNQCG RNFQGTSDLI RHQV Highest antigen sequence identity to the following orthologs - mouse 52%, rat 52%.
May be involved in transcriptional regulation.
Specifications
| ZNF594 | |
| Polyclonal | |
| Unconjugated | |
| ZNF594 | |
| KIAA1871; Zinc finger protein 594; zinc finger protein HZF18; ZNF594 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 84622 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunocytochemistry | |
| 0.1 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q96JF6 | |
| ZNF594 | |
| Recombinant Human ZNF594. Recombinant protein control fragment (Product #RP-107551). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction