missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ ZNF598 Monoclonal Antibody (K92040_5C9), Invitrogen™

Product Code. 30201363
Change view
Click to view available options
Quantity:
50 μL
200 μL
Unit Size:
200µg
50µg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30201363 200 μL 200µg
30207216 50 μL 50µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30201363 Supplier Invitrogen™ Supplier No. MA556099

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

Sequence of this protein is as follows: PRHSRRNEGV VGGEDYEEVD RYSRQGRVAR AGTRGAQQSR RGSWRYKREE EDREVAAAVR ASVAAQQQEE ARRSEDQEEG GRPKKEEAAA RGPEDPRGPR RSPRTQGEGP GPKETSTNGP VSQEAFSVTG

ZNF598 (E3 ubiquitin-protein ligase ZNF598) plays a key role in the ribosome quality control (RQC). RQC is a pathway that takes place when a ribosome has stalled during translation. It is required for ribosomes to terminally stall during translation of poly(A) sequences by mediating monoubiquitination of 40A ribosomal protein RPS10/eS10 and RPS3/uS3. Stalling precludes synthesis of a long poly-lysine tail and initiated the RQC pathway to degrade the potentially detrimental aberrant nascent polypeptide.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ZNF598
Applications Immunoprecipitation, Western Blot
Classification Monoclonal
Clone K92040_5C9
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol, 0.5% BSA and 0.09% sodium azide
Gene ZNF598
Gene Accession No. Q86UK7
Gene Alias BC023040; C6orf149; E3 ubiquitin-protein ligase ZNF598; FLJ00086; ISD11; Ntrap; Zfp598; Zinc finger protein 598; ZNF598
Gene Symbols ZNF598
Host Species Mouse
Immunogen Recombinant human ZNF598 (immunogen range is 291-420).
Purification Method Protein A
Quantity 200 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 90850
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.