missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ZNF599 Antibody, Alexa Fluor™ 700, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-98118AF700
This item is not returnable.
View return policy
Description
ZNF599 Polyclonal antibody specifically detects ZNF599 in Human samples. It is validated for Immunohistochemistry (Paraffin)
Specifications
| ZNF599 | |
| Polyclonal | |
| 50mM Sodium Borate | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| 148103 | |
| Store at 4°C in the dark. | |
| IgG |
| Immunohistochemistry (Paraffin) | |
| Alexa Fluor 700 | |
| FLJ30663, FLJ50920, zinc finger protein 599 | |
| Produced in rabbits immunized with E. coli-derived Human ZNF599 fragment. | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction