Learn More
ZNF611 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 17249-1-AP-20UL
This item is not returnable.
View return policy
Description
May be involved in transcriptional regulation.Specifications
| ZNF611 | |
| Polyclonal | |
| Unconjugated | |
| ZNF611 | |
| zinc finger protein 611, ZNF611 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 81856 | |
| -20°C | |
| Liquid |
| Western Blot | |
| 0.22 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q8N823 | |
| ZNF611 | |
| ZNF611 Fusion Protein Ag9984 | |
| 20 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |