missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ ZNF618 Polyclonal Antibody
Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA566417
This item is not returnable.
View return policy
Description
Immunogen sequence: CFQEHRDLHAV DVFSVEGAPE NRADPFDQGV VATDEVKEEP PEPFQKIGPK TGNYTCEFCG KQYKYYTPYQ EHVALHAPIS TAPGWEPPDD PDTGSECS Highest antigen sequence identity to the following orthologs - mouse 99%, rat 98%.
May be involved in transcriptional regulation.
Specifications
| ZNF618 | |
| Polyclonal | |
| Unconjugated | |
| ZNF618 | |
| FP13169; KIAA1952; NEDD10; neural precursor cell expressed, developmentally down-regulated 10; zinc finger protein 618; ZNF618 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 114991 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Western Blot, Immunocytochemistry | |
| 0.6 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q5T7W0 | |
| ZNF618 | |
| Recombinant Human ZNF618. Recombinant protein control fragment (Product #RP-105205). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction