missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ZNF621 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
€ 415.00 - € 624.00
Specifications
| Antigen | ZNF621 |
|---|---|
| Applications | Immunocytochemistry, Immunofluorescence |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Host Species | Rabbit |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18261095
|
Novus Biologicals
NBP2-56492 |
100 μL |
€ 624.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18625597
|
Novus Biologicals
NBP2-56492-25ul |
25 μL |
€ 415.00
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
ZNF621 Polyclonal specifically detects ZNF621 in Human samples. It is validated for Immunocytochemistry/Immunofluorescence.Specifications
| ZNF621 | |
| Polyclonal | |
| Rabbit | |
| Human | |
| DKFZp686B11107, FLJ23786, FLJ45246, MGC126668, zinc finger protein 621 | |
| ZNF621 | |
| IgG | |
| Affinity Purified |
| Immunocytochemistry, Immunofluorescence | |
| Unconjugated | |
| RUO | |
| PBS, pH 7.2, containing 40% glycerol with 0.02% Sodium Azide | |
| 285268 | |
| This antibody was developed against a recombinant protein corresponding to the following amino acid sequence:RGISQGGESWIKNEGLVIKQEASEETELHRMPVGGLLRNVSQHFDFKRKALKQTFNLNPNL | |
| Primary | |
| Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
For Research Use Only
Hittar du en möjlighet till förbättring?Dela en innehållskorrigering
Korrigering av produktinnehåll
Din input är viktig för oss. Fyll i det här formuläret för att ge feedback relaterad till innehållet på denna produkt.
Produkttitel