missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ZNF622 Antibody, Alexa Fluor™ 594, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-32102AF594
This item is not returnable.
View return policy
Description
ZNF622 Polyclonal antibody specifically detects ZNF622 in Human samples. It is validated for Western Blot
Specifications
| ZNF622 | |
| Polyclonal | |
| Western Blot | |
| MGC2485, zinc finger protein 622, Zinc finger-like protein 9, ZPR9MGC17552 | |
| The immunogen this antibody was made to, maps to a region between residue 125 to 175 of human Zinc Finger Protein 622 using the numbering given in entry NP_219482.1 (GeneID 90441). | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
| Western Blot | |
| Alexa Fluor 594 | |
| 50mM Sodium Borate | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| 90441 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction