missing translation for 'onlineSavingsMsg'
Learn More

ZNF622 Antibody (PCRP-ZNF622-1C11), DyLight 405, Novus Biologicals™

Product Code. 18502750 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
100 μg
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18502750 100 μg 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18502750 Supplier Novus Biologicals Supplier No. NBP308811V

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

ZNF622 Monoclonal antibody specifically detects ZNF622 in Human samples. It is validated for Flow Cytometry, Immunocytochemistry/ Immunofluorescence, Protein Array
TRUSTED_SUSTAINABILITY

Specifications

Antigen ZNF622
Applications Flow Cytometry, Immunofluorescence, Protein Array
Classification Monoclonal
Clone PCRP-ZNF622-1C11
Conjugate DyLight 405
Formulation 50mM Sodium Borate
Gene Alias MGC2485, zinc finger protein 622, Zinc finger-like protein 9, ZPR9MGC17552
Host Species Mouse
Immunogen Recombinant full-length human ZNF622 protein (Uniprot: Q969S3)
Purification Method Protein A or G purified
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 90441
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.