missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ ZNF622 Monoclonal Antibody (208CT34.1.1)

Mouse Monoclonal Antibody
Brand: Invitrogen™ MA537502
This item is not returnable.
View return policy
Description
ZNF622 Monoclonal Antibody for Western Blot
ZNF622 may behave as an activator of the bound transcription factor, MYBL2, and be involved in embryonic development.
Specifications
| ZNF622 | |
| Monoclonal | |
| 0.5 mg/mL | |
| PBS with 0.09% sodium azide; pH 7.4 | |
| Q91VY9 | |
| Zfp622 | |
| ZNF622 recombinant protein. | |
| 400 μL | |
| Primary | |
| Mouse | |
| Antibody | |
| IgG1 κ |
| Western Blot | |
| 208CT34.1.1 | |
| Unconjugated | |
| Zfp622 | |
| 1110033B05Rik; D15Ertd806e; liver regeneration-related protein LRRG121; Zfp622; Zinc finger protein 622; zinc finger-like protein 9; Znf622; ZPR9 | |
| Mouse | |
| Protein G | |
| RUO | |
| 52521 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction