missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IRF5 Antibody (903430) [Allophycocyanin], Novus Biologicals™
Rat Monoclonal Antibody
Brand: Novus Biologicals FAB8447A
This item is not returnable.
View return policy
Description
IRF5 Monoclonal antibody specifically detects IRF5 in Mouse samples. It is validated for Flow Cytometry (Intracellular Staining), CyTOF
Specifications
| IRF5 | |
| Monoclonal | |
| APC | |
| PBS | |
| Rat | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| Primary | |
| Mouse | |
| Purified |
| Flow Cytometry (Intracellular Staining), CyTOF | |
| 903430 | |
| Intracellular Staining by Flow Cytometry, CyTOF-ready | |
| IBD14, interferon regulatory factor 5, IRF-5, SLEB10 | |
| E. coli-derived recombinant mouse IRF5, Ala208-Phe288, Accession # P56477 | |
| 0.1 mL | |
| Apoptosis, Death Receptor Signaling Pathway | |
| 3663 | |
| Store at 4°C in the dark. | |
| IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction